missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate dihydrate, Puriss., meets analytical spec. of NF, E 516, 99.0-101.0% (based on anhydrous substance), Solstice

Catalog No. 6001922
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001922 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001922 Supplier Solstice Supplier No. 120561KG

missing translation for 'validMainframeMsg'

Puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
IUPAC Name calcium;sulfate;dihydrate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 kg
Linear Formula CaSO4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 172.17g/mol
Percent Purity 99 to 101% (based on anhydrous substance)
Grade puriss.
Chemical Name or Material Calcium Sulfate Dihydrate, Meets analytical specification of NF
EINECSNumber 231-900-3
RTECSNumber EW4150000