missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate, Dihydrate, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18609538
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609538 50 kg Poly Bottle
18609534 125 g Poly Bottle
18609535 500 g Poly Bottle
18609536 2.5 kg Poly Bottle
18609537 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609538 Supplier Spectrum Chemical Mfg Cor Supplier No. C116150KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
IUPAC Name calcium dihydrate sulfate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 50 kg
Percent Purity 98 to 102%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Calcium Sulfate, Dihydrate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.