missing translation for 'onlineSavingsMsg'
Learn More

Calcium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLCA23X

 View more versions of this product

Catalog No. PLCA2-3X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ca10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Calcium Standard
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00010897 MFCD00085314
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
OYPRJOBELJOOCE-UHFFFAOYSA-N
5% HNO3
40.08
5460341
Assurance
Triple-leached LDPE bottle
7440-70-2 , 7697-37-2 , 7732-18-5
Ca
AA, ICP
Certified Reference Material
[Ca]
calcium
500 mL
CHEBI:29320

Chemical Identifiers

7440-70-2
40.08
OYPRJOBELJOOCE-UHFFFAOYSA-N
CHEBI:29320
[Ca]
Ca
MFCD00010897 MFCD00085314
5460341
calcium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS Documents

Documents

Promotions

Promotions

Product intended for laboratory use only