missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Standard, 1000 ppm Ca, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Concentration or Composition Notes | 1,000ppm Ca |
| CAS Min % | 97.0, 0.37, 0.24 |
| CAS Max % | 100.0, 0.38, 0.25 |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Quantity | 500 mL |
| Chemical Name or Material | Calcium Standard Solution, Stock Solution |