missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Phosphate Tribasic, MP Biomedicals
Specifications
Specifications
| Quantity | 25 g |
| Formula Weight | 502.3 |
| Chemical Name or Material | Calcium Phosphate Tribasic |
Safety and Handling
| Recommended Storage | Room Temperature |