missing translation for 'onlineSavingsMsg'
Learn More

Calcium nitrate tetrahydrate, 99+%, ACS reagent

Catalog No. AC423535000
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423535000 500 g Plastic bottle
AC423530250 25 g Plastic bottle
AC423530025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC423535000 Supplier Thermo Scientific Chemicals Supplier No. 423535000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13477-34-4 | CaH8N2O10 | 236.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
MDL Number MFCD00149604
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
PubChem CID 16211656
ChEBI CHEBI:86159
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Specifications

Boiling Point 132.0°C
Melting Point 45.0°C
Color White
pH 5.0 to 7.0 (5% soln. at 25°C)
Physical Form Crystals
Quantity 500 g
Packaging Plastic bottle
Specific Gravity 1.86
Assay Percent Range 99.0 to 103% (Complexometry)
Density 1.8600g/mL
Linear Formula Ca(NO3)2·4H2O
Merck Index 15, 1684
Solubility Information Solubility in water: 1470g/L (0°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 236.15
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Calcium nitrate tetrahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May intensify fire; oxidizer.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
RTECSNumber EW3000000

RUO – Research Use Only