missing translation for 'onlineSavingsMsg'
Learn More

Calcium ICP Standard, 1000 ppm Ca in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PCA1KN500

 View more versions of this product

Catalog No. PCA1KN-500


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Calcium ICP Standard
471-34-1 , 7697-37-2 , 7732-18-5
93.0,4.65,0.23
1 mL = 1 mg Ca
CCaO3
Colorless
Approximately 0°C
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
500 mL
CHEBI:3311
Natural Poly Bottle
1,000ppm Ca
97.0,4.84,0.24
Liquid
MFCD00010906
< 2
Standard
[Ca++].[O-]C([O-])=O
100.09
10112

Chemical Identifiers

471-34-1
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
CHEBI:3311
[Ca++].[O-]C([O-])=O
CCaO3
MFCD00010906
10112
calcium carbonate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions