missing translation for 'onlineSavingsMsg'
Learn More

Calcium ICP Standard, 1000 ppm Ca in 3% HNO3, Ricca Chemical

Catalog No. PCA1KN100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PCA1KN100 100 mL Natural LDPE Bottle
PCA1KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PCA1KN100 Supplier Ricca Chemical Company Supplier No. PCA1KN100
Only null left

Chemical Identifiers

CAS 471-34-1 , 7697-37-2 , 7732-18-5
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Ca
CAS Min % 93.0, 4.65, 0.23
CAS Max % 97.0, 4.84, 0.24
Equivalence 1 mL = 1 mg Ca
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Chemical Name or Material Calcium ICP Standard
Show More Show Less