missing translation for 'onlineSavingsMsg'
Learn More

Calcium ICP Standard, 10,000 ppm Ca in 5% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PCA10KN500

 View more versions of this product

Catalog No. PCA10KN-500


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Calcium ICP Standard
471-34-1 , 7697-37-2 , 7732-18-5
85.0,10.52,2.13
1 mL = 10 mg Ca
CCaO3
Colorless
Standard
[Ca++].[O-]C([O-])=O
100.09
10112
Natural Poly Bottle
10,000ppm Ca
89.0,10.95,2.22
Liquid
MFCD00010906
< 2
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate
500 mL
CHEBI:3311

Chemical Identifiers

471-34-1
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
CHEBI:3311
[Ca++].[O-]C([O-])=O
CCaO3
MFCD00010906
10112
calcium carbonate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions