missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium ICP Standard, 10,000 ppm Ca in 5% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PCA10KN100
Specifications
| Calcium ICP Standard | |
| 471-34-1 , 7697-37-2 , 7732-18-5 | |
| 85.0,10.52,2.13 | |
| 1 mL = 10 mg Ca | |
| CCaO3 | |
| Colorless | |
| Standard | |
| [Ca++].[O-]C([O-])=O | |
| 100.09 | |
| 10112 |
| Natural LDPE Bottle | |
| 10,000ppm Ca | |
| 89.0,10.95,2.22 | |
| Liquid | |
| MFCD00010906 | |
| < 2 | |
| VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
| calcium carbonate | |
| 100 mL | |
| CHEBI:3311 |
Chemical Identifiers
| 471-34-1 | |
| 100.09 | |
| VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
| CHEBI:3311 | |
| [Ca++].[O-]C([O-])=O |
| CCaO3 | |
| MFCD00010906 | |
| 10112 | |
| calcium carbonate |