missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium ICP Standard, 10,000 ppm Ca in 5% HCl, Ricca Chemical
Supplier: Ricca Chemical Company PCA10KH500
Specifications
| Calcium ICP Standard | |
| 471-34-1 , 7647-01-0 , 7732-18-5 | |
| 90.0,9.77,2.25 | |
| CCaO3 | |
| Colorless | |
| Approximately 100°C | |
| Miscible with water | |
| Standard | |
| [Ca++].[O-]C([O-])=O | |
| 100.09 | |
| 10112 | |
| Laboratory |
| Natural Poly Bottle | |
| 86.0,9.39,2.16 | |
| Odorless | |
| MFCD00010906 | |
| < 2 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Calibration Standard | |
| VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
| calcium carbonate | |
| 500 mL | |
| CHEBI:3311 | |
| 1mL = 10mg Ca (10, 000ppm Ca)CaCO3 in 5% HCl |
Chemical Identifiers
| 471-34-1 | |
| 100.09 | |
| VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
| CHEBI:3311 | |
| [Ca++].[O-]C([O-])=O |
| CCaO3 | |
| MFCD00010906 | |
| 10112 | |
| calcium carbonate |
Safety and Handling
ShelfLife : 18 months