missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, Powder, FCC, 95-100.5%, Spectrum™ Chemical

Catalog No. 18601319
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601319 45 kg Poly Bottle
18601316 500 g Poly Bottle
18601317 2.5 kg Poly Bottle
18601318 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601319 Supplier Spectrum Chemical Mfg Cor Supplier No. C111645KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C1116, 1305-62-0, Ca(OH)2

Spectrum™ Chemical Calcium Hydroxide, Powder, FCC is used extensively in the food industry to clarify solutions, as a digestion aid, baking soda substitute and as a nutritional supplement because of its low toxicity. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
IUPAC Name calcium dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

Physical Form Crystalline Powder or Needles
CAS Max % 100%
Quantity 45 kg
Packaging Poly Bottle
Percent Purity 95 to 100.5%
Grade FCC
Chemical Name or Material Calcium Hydroxide