missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydride, 98% (metals basis excluding Mg), Mg <1%

Catalog No. AA4164530
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4164530 250 g
AA4164518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4164530 Supplier Thermo Scientific Chemicals Supplier No. 04164530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7789-78-8 | CaH2

Solvent in Sol-Gel precursor applications. Calcium hydride is used as an efficient drying agent for aprotic base-stable solvents like ethers and tertiary amines. It is a useful dehydrating agent in a synthesis of aldehyde enamines in high yield and purity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-78-8
Molecular Formula CaH2
Molecular Weight (g/mol) 42.09
MDL Number MFCD00010897
InChI Key FAQLAUHZSGTTLN-UHFFFAOYSA-N
IUPAC Name calcium dihydride
SMILES [CaH2]

Specifications

Melting Point 600°C (decomposition)
Color Gray
Physical Form Powder
Quantity 250 g
Assay Percent Range 98%
Solubility Information Soluble in water and alcohol. Insoluble in benzene.
Formula Weight 42.1
Sensitivity Moisture sensitive
Appearance Light gray
Density 1.9
Chemical Name or Material Calcium hydride, Mg <1%
Show More Show Less
Hazard Category H260-H315-H319
Precautionary Statement P223-P231+P232-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P335+P334-P362-P370+P378q-P501c
DOTInformation DOT Class: 4.3, Packaging Group: I
EINECSNumber 232-189-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only