missing translation for 'onlineSavingsMsg'
Learn More

Medisca Calcium Citrate, USP

Catalog No. 14186161
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186161 500 g
14186062 100 g
14186064 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14186161 Supplier Medisca Supplier No. 108008*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Calcium Citrate, USP is a white, odorless, crystalline powder that is slightly hygroscopic.

Calcium Citrate, USP is a white, odorless, crystalline powder that is slightly hygroscopic.

Alternative name(s): Tricalcium citrate

Specifications

Chemical Name or Material Calcium Citrate, USP
Packaging Jar, 1000mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 5785-44-4
Type Drug Standard
Molecular Weight (g/mol) 570.49
Quantity 500 g
Percent Purity 98 to 102 %
Grade USP