missing translation for 'onlineSavingsMsg'
Learn More

Calcium citrate tetrahydrate, 96%

Catalog No. AAA1482822
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1482822 100 g
AAA1482836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1482822 Supplier Thermo Scientific Chemicals Supplier No. A1482822

CAS: 5785-44-4 | C12H18Ca3O18 | 570.492 g/mol

Calcium citrate tetrahydrate may be used as raw material in the preparation of nanoparticle with the nominal composition Ca0.8 Ba0.2 Ti03 :Pr3+, via sol-gel process. It may be employed as carbon source in a simple and effective carbonization method to prepare porous carbon without any activation process. It is suitable for use in the preparation of bovine serum albumin (BSA) nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium citrate tetrahydrate may be used as raw material in the preparation of nanoparticle with the nominal composition Ca0.8 Ba0.2 Ti03 :Pr3+, via sol-gel process. It may be employed as carbon source in a simple and effective carbonization method to prepare porous carbon without any activation process. It is suitable for use in the preparation of bovine serum albumin (BSA) nanoparticles.

Solubility
Soluble in water and hydrochloric acid.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 5785-44-4
Molecular Formula C12H18Ca3O18
Molecular Weight (g/mol) 570.492
MDL Number MFCD00150786
InChI Key LNIZKKFWMDARJV-UHFFFAOYSA-H
Synonym calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f
PubChem CID 5282392
IUPAC Name tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2]

Specifications

Quantity 100 g
Solubility Information Soluble in water and hydrochloric acid.
Formula Weight 572.60 (500.54 Anhydrous)
Percent Purity 96%
Chemical Name or Material Calcium citrate tetrahydrate
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only