missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride Standard, 1000 ppm CaCO3 (400 ppm Ca), 0.0100 M Ca2+, 0.0200 N Ca2+, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 98.0, 0.19, 0.1 |
| CAS Max % | 100.0, 0.2, 0.1 |
| Equivalence | 1mL= 1 mg CaCO3 |
| Boiling Point | 100°C |
| Quantity | 120 mL |
| Concentration or Composition (by Analyte or Components) | 1000ppm CaCO3 (400ppm Ca), 0.0100 M Ca2+, 0.0200 N Ca2+ |
| Chemical Name or Material | Calcium Chloride Standard |