missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Solution, Fisher Chemical

Catalog No. SC101
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SC101 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SC101 Supplier Thermo Fisher Scientific Supplier No. SC10 1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For biochemical oxygen demand

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Chemical Identifiers

CAS 10043-52-4 , 7732-18-5
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00149613
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Color Undesignated
Melting Point 0°C
Concentration 2.5 to 3.0%
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 14mmHg at 16°C
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Calcium Chloride Solution