missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calcium Chloride, Dihydrate, Calbiochem™,

Catalog No. 2082901KG
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2082901KG 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2082901KG Supplier MilliporeSigma™ Supplier No. 2082901KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hygroscopic

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·2(H2O)
Synonym CaCl2

Specifications

Quantity 250 g
Assay Percent Range ≥99%
Solubility Information Easily soluble in cold water
Formula Weight 147
Chemical Name or Material Calcium Chloride, Dihydrate
Grade by complexometry
RTECSNumber EV9810000
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).