missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, Desiccant, ACS, 96%, Spectrum™ Chemical
$197.09 - $14761.59
Chemical Identifiers
| CAS | 10043-52-4 |
|---|---|
| Molecular Formula | CaCl2 |
| Molecular Weight (g/mol) | 110.98 |
| InChI Key | UXVMQQNJUSDDNG-UHFFFAOYSA-L |
| IUPAC Name | calcium dichloride |
| SMILES | [Cl-].[Cl-].[Ca++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18609470
|
Spectrum Chemical Mfg Cor
C153625GM |
25 g | Poly Bottle |
Each for $197.09
|
|
||||
|
18609471
|
Spectrum Chemical Mfg Cor
C1536100GM |
100 g | Poly Bottle |
Each for $364.00
|
|
||||
|
18609472
|
Spectrum Chemical Mfg Cor
C1536500GM |
500 g | Poly Bottle |
Each for $795.38
|
|
||||
|
18609473
|
Spectrum Chemical Mfg Cor
C153625KG |
2.5 kg | Poly Bottle |
Each for $1,847.47
|
|
||||
|
18609474
|
Spectrum Chemical Mfg Cor
C153612KG |
12 kg | Poly Pail |
Each for $6,091.26
|
|
||||
|
18609475
|
Spectrum Chemical Mfg Cor
C153650KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 10043-52-4 | |
| 110.98 | |
| calcium dichloride |
| CaCl2 | |
| UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
| [Cl-].[Cl-].[Ca++] |
Specifications
| 10043-52-4 | |
| 25 g | |
| UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
| calcium dichloride | |
| 96% | |
| Poly Bottle |
| 1 | |
| CaCl2 | |
| [Cl-].[Cl-].[Ca++] | |
| 110.98 | |
| ACS |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.