missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride, Desiccant, ACS, 96%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor C1536100GM
Specifications
| 10043-52-4 | |
| 100 g | |
| UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
| calcium dichloride | |
| 96% | |
| Poly Bottle |
| 1 | |
| CaCl2 | |
| [Cl-].[Cl-].[Ca++] | |
| 110.98 | |
| ACS |
Chemical Identifiers
| 10043-52-4 | |
| 110.98 | |
| calcium dichloride |
| CaCl2 | |
| UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
| [Cl-].[Cl-].[Ca++] |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.