missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, Desiccant, ACS, 96%, Spectrum™ Chemical

Catalog No. 18609470
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609470 25 g Poly Bottle
18609471 100 g Poly Bottle
18609472 500 g Poly Bottle
18609473 2.5 kg Poly Bottle
18609474 12 kg Poly Pail
18609475 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18609470 Supplier Spectrum Chemical Mfg Cor Supplier No. C153625GM

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 96%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.