missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorure de calcium (Dessiccant, 20 mailles et plus fin) Grade ACS, Fisher Chemical™
Spécifications
Spécifications
| Nom ou substance chimique | Calcium Chloride |
| Nom Remarque | Desséchant |
| Point d’ébullition | 1600 °C |
| pH | 8 à 10 |
| Point de fusion | 782 °C |
| Forme physique | Solide |
| Couleur | Beige |
| Densité | 2.15g/mL |
| Matériau | Chlorure de calcium |
| Quantité | 500 g |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Irritating to eyes. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention. NFPA Health:2 Flammability:0 Instability:1 |