missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, 1M aq. soln.

Catalog No. AAJ63122AE
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63122AE 100 mL
AAJ63122AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63122AE Supplier Thermo Scientific Chemicals Supplier No. J63122AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10043-52-4 | CaCl2 | 110.98 g/mol

Calcium chloride is used as a de-icing and dust control of roads, drilling muds, dust proofing, freeze proofing and thawing coal, coke, stone, sand, and ore, concrete conditioning, paper and pulp industry, drying and desiccating agent, and tire weighting. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability. Calcium chloride is a soluble compound that can serve as a source of calcium ions in a solution. Can be useful for displacing ions from solution, for example, phosphate may be displaced from solution by calcium. Calcium chloride solution has many applications in cell and molecular biology experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium chloride is used as a de-icing and dust control of roads, drilling muds, dust proofing, freeze proofing and thawing coal, coke, stone, sand, and ore, concrete conditioning, paper and pulp industry, drying and desiccating agent, and tire weighting. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability. Calcium chloride is a soluble compound that can serve as a source of calcium ions in a solution. Can be useful for displacing ions from solution, for example, phosphate may be displaced from solution by calcium. Calcium chloride solution has many applications in cell and molecular biology experiments.

Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong acids, borane/boron oxides, zinc, calcium oxide, methyl vinyl ether. Calcium chloride is attacked by bromine trifluoride. Anhydrous Calcium Chloride is highly hygroscopic and is often used as a desiccant.

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Color Colorless
Physical Form Liquid
Quantity 100 mL
Solubility Information Soluble in water (74.5g/100ml) at 20°C,alcohol,water (159g/100ml) at 100°C,acetic acid,and acetone.
Formula Weight 110.99
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Odor Odorless
Chemical Name or Material Calcium chloride
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only