missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Carbonate Standard , (1mL = 1mg CaCO3 = 0.4mg Ca), for Hardness, Certified, 1.000 ±0.010mg CaCO3/mL, LabChem™
Description
Calcium Carbonate Standard, CCaO3, CAS Number-471-34-1, 7647-01-0, 500mL, 98.9, 1, 0.1, CHEBI:3311, Colorless, 1.000 +/- 0.010mg CaCO3/mL, 1g/mL, 100.09, Certified, Warning, GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Specifications
Specifications
| Packaging | Poly Bottle |
| CAS Max % | 98.9, 1, 0.1 |
| Identification | Passes Test |
| Density | 1g/mL |
| Physical Form | Liquid |
| Concentration | 1.000 ±0.010mg CaCO3/mL |
| Linear Formula | CaCO3 |
| Color | Colorless |
| Solubility | Soluble in water |
| Quantity | 125 mL |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. Take off contaminated clothing. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| Recommended Storage | Room Temperature |