missing translation for 'onlineSavingsMsg'
Learn More

Calcium Carbonate, Powder, Low in Alkalies, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02002517
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002517 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02002517 Supplier Avantor Supplier No. 129401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 471-34-1
Physical Form Powder
Quantity 500 g
Molecular Formula CCaO3
MDL Number MFCD00010906
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES [Ca++].[O-]C([O-])=O
IUPAC Name calcium;carbonate
Molecular Weight (g/mol) 100.09
PubChem CID 10112
ChEBI CHEBI:3311
Formula Weight 100.09
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Chemical Name or Material Calcium Carbonate
Show More Show Less