missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014238
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014238 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014238 Supplier Merck Emd Millipore Supplier No. PHR13265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Calcium carbonate is a material that is widely found in nature, and is commonly referred to as calcite. Calcium carbonate is used in such processes as the manufacture of paper, paints, glass, and ceramics, the compounding of polymers, environmental applications such as wastewater treatment, and as an extender in paints and coatings. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 471-34-1
Density 2.93 g/mL (at 25°C)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula CaCO3
MDL Number MFCD00010906
Synonym Calcium carbonate
Type Certified Reference Material
Quantity 5 g
Formula Weight 100.09
Grade Certified Reference Material
Show More Show Less