missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Carbonate, Light Powder, U.S.P. - F.C.C., J.T. Baker™
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 500 g |
| IUPAC Name | calcium;carbonate |
| Formula Weight | 100.09 |
| Grade | FCC/USP |
| Packaging | Poly Bottle |
| Chemical Name or Material | Calcium Carbonate |