missing translation for 'onlineSavingsMsg'
Learn More

Calcium Bis(fluorosulfonyl)imide 98.0+%, TCI America™

Numéro de catalogue. C34761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C34761G 1 g
C34765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C34761G Fournisseur TCI America Code fournisseur C34761G
Il en reste null

Identifiants chimiques

CAS 1906900-37-5
Molecular Formula CaF4N2O8S4

Spécifications

Color White
Quantity 1 g
UN Number 1759
Formula Weight 400.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium Bis(fluorosulfonyl)imide