missing translation for 'onlineSavingsMsg'
Learn More

Calcium arsenate, 95%

Catalog No. AA3921930
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Packing group II
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA3921930 250 g Packing group II
AA3921918 50 g Packing group II
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3921930 Supplier Thermo Scientific Chemicals Supplier No. 03921930

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7778-44-1 | As2Ca3O8 | 398.069 g/mol

Calcium arsenate is also used in weather- resistant wood treatment. It was used mainly in viticulture, in the past. it was mainly used against the boll weevil in cotton cultivation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7778-44-1
Molecular Formula As2Ca3O8
Molecular Weight (g/mol) 398.069
MDL Number MFCD00049407
InChI Key RMBBSOLAGVEUSI-UHFFFAOYSA-H
Synonym calcium arsenate, pencal, cucumber dust, spra-cal, calcium-o-arsenate, chip-cal granular, tricalcium arsenate, calcium orthoarsenate, kalziumarseniat german, arsenic acid, calcium salt
PubChem CID 24501
IUPAC Name tricalcium;trioxido(oxo)-$l^{5}-arsane
SMILES [O-][As](=O)([O-])[O-].[O-][As](=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2]

Specifications

Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 95%
Solubility Information It is soluble in water and acids,insoluble in organic solvents
Formula Weight 398.03
Packaging Packing group II
Density 3.62 g/cm3 at 20°C
Chemical Name or Material Calcium arsenate
Hazard Category H301+H331-H350
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 231-904-5
RTECSNumber CG0830000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only