missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium AA Standard, 1000 ppm Ca in 3% HCl, Ricca Chemical
$112.43 - $112.43
Chemical Identifiers
| CAS | 471-34-1 |
|---|---|
| Molecular Formula | CCaO3 |
| Molecular Weight (g/mol) | 100.09 |
| MDL Number | MFCD00010906 |
| InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
| PubChem CID | 10112 |
| ChEBI | CHEBI:3311 |
| IUPAC Name | calcium carbonate |
| SMILES | [Ca++].[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
ACA1KH-100
|
Ricca Chemical Company
ACA1KH100 |
100 mL | Natural LDPE Bottle |
Each for $112.43
|
|
||||
Chemical Identifiers
| 471-34-1 | |
| 100.09 | |
| VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
| CHEBI:3311 | |
| [Ca++].[O-]C([O-])=O |
| CCaO3 | |
| MFCD00010906 | |
| 10112 | |
| calcium carbonate |
Specifications
| Natural LDPE Bottle | |
| 1,000ppm Ca | |
| 98.0,3.95,0.24 | |
| Liquid | |
| MFCD00010906 | |
| 100°C | |
| [Ca++].[O-]C([O-])=O | |
| 100.09 | |
| 10112 | |
| Calcium AA Standard |