missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium AA Standard, 1000 ppm Ca in 3% HCl, Ricca Chemical
Supplier: Ricca Chemical Company ACA1KH100
Specifications
| Calcium AA Standard | |
| 471-34-1 , 7647-01-0 , 471-34-1 | |
| 94.0,3.79,0.23 | |
| 1 mL = 1 mg Ca | |
| CCaO3 | |
| Colorless | |
| VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
| calcium carbonate | |
| 100 mL | |
| CHEBI:3311 |
| Natural LDPE Bottle | |
| 1,000ppm Ca | |
| 98.0,3.95,0.24 | |
| Liquid | |
| MFCD00010906 | |
| 100°C | |
| [Ca++].[O-]C([O-])=O | |
| 100.09 | |
| 10112 |
Chemical Identifiers
| 471-34-1 | |
| 100.09 | |
| VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
| CHEBI:3311 | |
| [Ca++].[O-]C([O-])=O |
| CCaO3 | |
| MFCD00010906 | |
| 10112 | |
| calcium carbonate |