missing translation for 'onlineSavingsMsg'
Learn More

Calcein Blue 98.0+%, TCI America™

Catalog No. U00011G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00011G Supplier TCI America Supplier No. U00011G
Only null left

Chemical Identifiers

CAS 54375-47-2
Molecular Formula C15H15NO7
Molecular Weight (g/mol) 321.285
MDL Number MFCD00006867
InChI Key RJPSHDMGSVVHFA-UHFFFAOYSA-N
Synonym 4-Methylumbelliferonemethyleneiminodiacetic Acid, Umbelliferone Complexon
PubChem CID 5393133
IUPAC Name 2-[carboxymethyl-[(7-hydroxy-4-methyl-2-oxochromen-8-yl)methyl]amino]acetic acid
SMILES CC1=CC(=O)OC2=C1C=CC(=C2CN(CC(=O)O)CC(=O)O)O

Specifications

Formula Weight 321.29
Percent Purity ≥98.0% (T)
Color White/Yellow
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Calcein Blue