missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Wee1 Inhibitor II

Catalog No. 6816411MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6816411MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6816411MG Supplier MilliporeSigma™ Supplier No. 6816411MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Wee1 Inhibitor II, also referenced under CAS 622855-50-9, controls the biological activity of Wee1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas

Specifications

Color Brown
Description 97% by HPLC
Product Type Inhibitor
Target Proteins Wee1
CAS 622855-50-9
Molecular Formula C24H19ClN2O3
Formula Weight 418.9g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Show More Show Less