missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ VEGFR3 Kinase Inhibitor, MAZ51

Catalog No. 67649210MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67649210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67649210MG Supplier MilliporeSigma™ Supplier No. 67649210MG

The VEGFR3 Kinase Inhibitor, MAZ51, also referenced under CAS 163655-37-6, controls the biological activity of VEGFR3 Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Color Red
Description 95% by HPLC
Target Proteins VEGF receptor tyrosine kinase
CAS 163655-37-6
Molecular Formula C21H18N2O
Formula Weight 314.1g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg