missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Ubiquitin Aldehyde

Catalog No. 66205650UG
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66205650UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66205650UG Supplier MilliporeSigma™ Supplier No. 66205650UG

Potent and specific inhibitor of multiple ubiquitin hydrolases involved in pathways of intracellular protein modification and turnover.

Potent and specific inhibitor of multiple ubiquitin hydrolases involved in pathways of intracellular protein modification and turnover. Useful for stabilizing endogenous or in vitro synthesized ubiquitin-protein conjugates. May also be used to enhance or decrease rates of ubiquitin-dependent degradation.

Specifications

Formulation In 150 mM HCL.
Description 95% by HPLC
Target Proteins multiple ubiquitin hydrolases involved in pathways of intracellular protein modification and turnover
Formula Weight 8500g/mol
Solubility Aqueous Buffers or DMSO
Form Liquid
Quantity 50 μg