missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ TLR4 Inhibitor, TAK-242

Catalog No. 6143165MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6143165MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6143165MG Supplier MilliporeSigma™ Supplier No. 6143165MG

TAK-242; CAS 243984-11-4; is a cell-permeable compound that selectively binds to Cys747 of TLR4 and selectively disrupts its interaction with adaptor molecules TIRAP and TRAM.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach

Specifications

Color Yellowish White
Description 98% by HPLC
Target Proteins TLR4
CAS 243984-11-4
Molecular Formula C15H17ClFNO4S
Formula Weight 361.8g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg