missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ TLR3/dsRNA Complex Inhibitor

Catalog No. 61431010MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61431010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61431010MG Supplier MilliporeSigma™ Supplier No. 61431010MG

The TLR3/dsRNA Complex Inhibitor blocks dsRNA binding to TLR3.

Packaged Under Inert Gas

Specifications

Color White
Description 99% by HPLC
Molecular Formula C18H13ClFNO3S
Formula Weight 377.8g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg