missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Thymidine

Catalog No. 60605GM
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60605GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60605GM Supplier MilliporeSigma™ Supplier No. 60605GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For selection of hybridoma cell lines

  • Used in conjunction with hypoxanthine and aminopterin
  • Reported to induce cell death in L-cells

Specifications

Format White solid
Product Type Thymidine
Content And Storage 15°C to 30°C
Purity ≥98% by HPLC
Quantity 5 g

For Research Use Only