missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ TGF-β RI Kinase Inhibitor VI, SB431542

Catalog No. 6164615MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6164615MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6164615MG Supplier MilliporeSigma™ Supplier No. 6164615MG

TGF-β RI Kinase Inhibitor VI, SB431542, CAS 301836-41-9, is a cell-permeable inhibitor of SMAD2 phosphorylation. Inhibits the activity of ALK4 and ALK5 (IC50 = 140 nM and 94 nM, respectively).

Packaged Under Inert Gas

Specifications

Color Off-white
Description 97% by HPLC
CAS 301836-41-9
Molecular Formula C22H16N4O3·2H2O
Formula Weight 420.42g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg