missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ ADP-HPD, Dihydrate, Ammonium Salt

Catalog No. p-7060020
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
60 μg
Catalog No. Quantity
11841560UG 60 μg
1 options

Catalog No. 11841560UG

Supplier: MilliporeSigma™ 11841560UG

Only null left

ADP-HPD, Dihydrate is an amino analog of ADP-ribose that acts as a highly potent, noncompetitive, and specific inhibitor of poly(ADP-ribose) glycohydrolase (PARG; IC50 = 120 nM).

Packaged Under Inert Gas

Specifications

Color White
Target Proteins PARG
Molecular Formula C15H23N6O12P2·NH4·2H2O
Formula Weight 595.4g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Lyophilized
Quantity 60 μg
Purity 95% by HPLC