missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Adenosine 3',5'-cyclic Monophosphate, Sodium Salt
Description
A key regulator of many cellular reactions. Released in response to hormonal stimulation. An activator of protein kinase A. Directly activates Epac, a Rap1 guanine-nucleotide exchange factor. Acts as a second messenger in signal transduction mechanisms. Promotes apoptosis in resting human B lymphocytes.
Specifications
Specifications
| Color | White |
| Target Proteins | Protein kinase A |
| CAS | 37839-81-9 |
| Molecular Formula | C10H11N5O6P·Na |
| Formula Weight | 351.2g/mol |
| Solubility | H2O |
| Content And Storage | Hygroscopic |
| Form | Solid |
| Quantity | 100 mg |
| Purity | 98% by HPLC |