missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Adenosine 3',5'-cyclic Monophosphate, Sodium Salt

Catalog No. 1168011GM
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1168011GM 1 g
116801100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1168011GM Supplier MilliporeSigma™ Supplier No. 1168011GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A key regulator of many cellular reactions.

A key regulator of many cellular reactions. Released in response to hormonal stimulation. An activator of protein kinase A. Directly activates Epac, a Rap1 guanine-nucleotide exchange factor. Acts as a second messenger in signal transduction mechanisms. Promotes apoptosis in resting human B lymphocytes.

Specifications

Color White
Target Proteins Protein kinase A
CAS 37839-81-9
Molecular Formula C10H11N5O6P·Na
Formula Weight 351.2g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 1 g
Purity 98% by HPLC