missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ STING Agonist, 2ʹ3ʹ-cGAMP

Catalog No. 5318890001
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5318890001 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5318890001 Supplier MilliporeSigma™ Supplier No. 5.31889.0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A cyclic-dinucleotide (cyclic-GMP-AMP) based compound that binds to the C-terminal domain of adaptor endoplasmic reticulum protein known as STING with high affinity (Kd = 4nM) and induces the synthesis of interferon-b (IFN-b) in mammalian cells (EC50 =

A cyclic-dinucleotide (cyclic-GMP-AMP) based compound that binds to the C-terminal domain of adaptor endoplasmic reticulum protein known as STING with high affinity (Kd = 4nM) and induces the synthesis of interferon-b (IFN-b) in mammalian cells (EC50 = 20nM). Shown to increase IFN-b production in M2-polarized macrophages associated with non-small cell lung carcinoma and enhances the expression of M1 markers iNOS and IL-12p40, while simultaneously reducing the expression of the M2 markers, Arg-1 and Fizz1. Exhibits about 300 fold greater affinity for STING than c-DiGMP, 3′,2′-cGAMP, and 3′,3′-cGAMP and about 75 fold greater affinity over 2′,2′-cGAMP.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Specifications

Color White
Description 98% by HPLC
Target Proteins STING
Molecular Formula C20H24N10O13P2 (free acid)
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 500 μg