missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ STAT5 Inhibitor III, Pimozide

Catalog No. 573110100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
573110100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 573110100MG Supplier MilliporeSigma™ Supplier No. 573110100MG

The STAT5 Inhibitor III, Pimozide, also referenced under CAS 2062-78-4, controls the biological activity of STAT5.

Packaged Under Inert Gas

Specifications

Color White
Description 98% by HPLC
CAS 2062-78-4
Molecular Formula C28H29F2N3O
Formula Weight 461.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 100 mg