missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ STAT3 Inhibitor XIII, C188-9
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | Off-white to Pale Beige |
| Description | 98% by HPLC |
| Product Type | Inhibitor |
| Molecular Formula | C27H21NO5S |
| Formula Weight | 471.5g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Powder |
| Quantity | 10 mg |