missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ STAT3 Inhibitor III, WP1066

Catalog No. 57309710MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57309710MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57309710MG Supplier MilliporeSigma™ Supplier No. 57309710MG

STAT3 Inhibitor III, WP1066, CAS 857064-38-1, is a cell-permeable AG490 tyrphostin analog that acts as a STAT3 pathway inhibitor. A potent anti-tumor agent.

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Color Off-white
Description 97% by HPLC
Target Proteins STAT3 pathway
CAS 857064-38-1
Molecular Formula C17H14BrN3O
Formula Weight 356.2g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg