missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ SPUTOLYSIN™ Reagent

Catalog No. 5600001SET
Change view
Click to view available options
Quantity:
1 Set (20 x 10 mL)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5600001SET 1 Set (20 x 10 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5600001SET Supplier MilliporeSigma™ Supplier No. 5600001SET

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For isolation of epithelial cells, pathogenic or saprophytic bacteria, fungi, and yeasts from sputum

  • Concentrate of Dithiothreitol in phosphate buffer, pH 6.5-7.5
  • Intended for the isolation of epithelial cells, pathogenic or saprophytic bacteria, fungi, and yeasts from sputum
  • Each vial is sufficient to make 100mL of working solution

Specifications

CAS 12-3-3483
Content And Storage Store at +15°C to +30°C
Formulation 20 vials each containing 10mL of concentrated DTT OR 1 vial containing 10mL of concentrated DTT
Molecular Formula C4H10O2S2
Quantity 1 Set (20 x 10 mL)
Form Liquid