missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Sorbitol

Produced by reduction of glucose

Supplier:  MilliporeSigma™ 567551KG

Catalog No. 567551KG


Only null left
Explore available promotions

Description

Description

  • Sugar alcohol
  • CAS: 50-70-4
  • Molecular formula: C6H14O6
  • Color: White
Specifications

Specifications

1 kg
15°C to 30°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions