missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Sodium Orthovanadate, Activated

Catalog No. 5086050001
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5086050001 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5086050001 Supplier MilliporeSigma™ Supplier No. 5.08605.0001

A depolymerized vanadate prepared from Sodium Orthovanadate with enhanced ability to competitively inhibit endogenous phosphatases.

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach

Specifications

Color Colorless
Target Proteins Phosphatases
Formula Weight 183.9g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Protect from Light
Form Liquid
Quantity 1 Set