missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Smoothened Agonist, SAG

Catalog No. 5666605MG
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5666605MG 5 mg
5666601MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5666605MG Supplier MilliporeSigma™ Supplier No. 5666605MG

A cell-permeable Smoothened Agonist, SAG, CAS 364590-63-6, modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM).

Packaged Under Inert Gas

Specifications

Color Light Yellow
CAS 364590-63-6
Molecular Formula C28H28ClN3OS·2HCl·2H2O
Formula Weight 599g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Purity 98% by HPLC